Skip to Content
MilliporeSigma
Supel™ QuE Z-Sep+ with MgSO4 Tube, pk of 50, centrifuge tube volume 15 mL , Shaker Compatible

Supel QuE QuEChERS tube

Supel™ QuE Z-Sep+ with MgSO4 Tube, pk of 50, centrifuge tube volume 15 mL , Shaker Compatible
Synonym(s):
QuEChERS

Products

Product Category
Brand
Product Line
Agency
Chemical Composition
Separation Technique
Volume (ml)
Quality Segment
Application
Package Size
Technique
Showing 1-20 of 30
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2