Skip to Content
MilliporeSigma
Sulfur trioxide pyridine complex 97%

Sulfur trioxide pyridine complex

Sulfur trioxide pyridine complex 97%
Synonym(s):
NSC 75831, Pyridine sulfur trioxide complex, Sulfur trioxide pyridine complex
Empirical Formula (Hill Notation):
C5H5N · O3S
CAS Number:
Molecular Weight:
159.16

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reagent Type
Special Grade
Showing 1-3 of 3