Skip to Content
MilliporeSigma
Sulfuric acid-d2 solution 96-98 wt. % in D2O, 99.5 atom % D

Sulfuric acid-d2 solution

Sulfuric acid-d2 solution 96-98 wt. % in D2O, 99.5 atom % D
Synonym(s):
Dideuterosulfuric acid solution
Linear Formula:
D2SO4/D2O
CAS Number:
Molecular Weight:
100.09
EC Number:

Products

Product Category
Formula Weight
Physical Form
Isotope
Markush Class
Markush Group
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing