Skip to Content
MilliporeSigma
Sulfanilic acid azochromotrop ≥80%

Sulfanilic acid azochromotrop

Sulfanilic acid azochromotrop ≥80%
Synonym(s):
1,8-Dihydroxy-2-(4-sulfophenylazo)naphthalene-3,6-disulfonic acid trisodium salt, 2-(4-Sulfophenylazo)-1,8-dihydroxy-3,6-naphthalene disulfonic acid trisodium salt, 2-(4-Sulfophenylazo)chromotropic acid trisodium salt, SPADNS
Empirical Formula (Hill Notation):
C16H9N2Na3O11S3
CAS Number:
Molecular Weight:
570.41
EC Number:

Products

Product Category
Color
Formula Weight
Physical Form
Purity
Chemical Composition
Application
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing