Skip to Content
MilliporeSigma
Sulfaclozine sodium VETRANAL®, analytical standard

Sulfaclozine sodium

Sulfaclozine sodium VETRANAL®, analytical standard
Empirical Formula (Hill Notation):
C10H8ClN4NaO2S
CAS Number:
Molecular Weight:
306.70
EC Number:

Products

Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing