Skip to Content
MilliporeSigma
Sulcotrione PESTANAL®, analytical standard

Sulcotrione

Sulcotrione PESTANAL®, analytical standard
Synonym(s):
2-[2-Chloro-4-(methylsulfonyl)benzoyl]-1,3-cyclohexanedione
Empirical Formula (Hill Notation):
C14H13ClO5S
CAS Number:
Molecular Weight:
328.77

Products

Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing