Skip to Content
MilliporeSigma
Sudan Blue II Dye content 98 %

Sudan Blue II

Sudan Blue II Dye content 98 %
Synonym(s):
Solvent Blue 35
Empirical Formula (Hill Notation):
C22H26N2O2
CAS Number:
Molecular Weight:
350.45
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Chemical Composition
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing