Skip to Content
MilliporeSigma
Stevioside primary reference standard

Stevioside

Stevioside primary reference standard
Synonym(s):
(4α)-13-[(2-O-β-D-Glucopyranosyl-β-D-glucopyranosyl)oxy]kaur-16-en-18-oic acid β-D-glucopyranosyl ester, Steviosin
Empirical Formula (Hill Notation):
C38H60O18
CAS Number:
Molecular Weight:
804.87
EC Number:

Products

Formula Weight
Manufacturer
Physical Form
Product Line
Format
Application
API Family
Reference Material Type
Special Grade