Skip to Content
MilliporeSigma
Stearic acid-13C18 99 atom % 13C, 99% (CP)

Stearic acid-13C18

Stearic acid-13C18 99 atom % 13C, 99% (CP)
Synonym(s):
Octadecanoic acid-13C18, Stearic acid-ul-13C
Linear Formula:
13CH3(13CH2)1613CO2H
CAS Number:
Molecular Weight:
302.35

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Isotope
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing