Skip to Content
MilliporeSigma
(S)-(−)-Propylene oxide 99%

(S)-(−)-Propylene oxide

(S)-(−)-Propylene oxide 99%
Synonym(s):
(S)-(−)-1,2-Epoxypropane, (S)-(−)-Methyloxirane
Empirical Formula (Hill Notation):
C3H6O
CAS Number:
Molecular Weight:
58.08
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing