Skip to Content
MilliporeSigma
(S)-(−)-Propylene carbonate 98%

(S)-(−)-Propylene carbonate

(S)-(−)-Propylene carbonate 98%
Synonym(s):
(S)-1,2-Propanediol cyclic carbonate
Empirical Formula (Hill Notation):
C4H6O3
CAS Number:
Molecular Weight:
102.09

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing