![Spironolactone for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/217/271/3227737a-1b85-4d33-a146-7743d48ba487/640/3227737a-1b85-4d33-a146-7743d48ba487.png)
Spironolactone for system suitability
![Spironolactone for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/217/271/3227737a-1b85-4d33-a146-7743d48ba487/640/3227737a-1b85-4d33-a146-7743d48ba487.png)
Synonym(s):
Spironolactone, 4-Pregnen-21-oic acid-17α-ol-3-one-7α-thiol γ-lactone 7-acetate, 7α-(Acetylthio)-17α-hydroxy-3-oxopregn-4-ene-21-carboxylic acid γ-lactone
Empirical Formula (Hill Notation):
C24H32O4S
CAS Number:
Molecular Weight:
416.57
Products
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing