Skip to Content
MilliporeSigma
Soyasapogenol B phyproof® Reference Substance

Soyasapogenol B

Soyasapogenol B phyproof® Reference Substance
Synonym(s):
(3β,4β,22β)-Olean-12-ene-3,22,23-triol, Soyasapogenin B, Soyasapogenol I
Empirical Formula (Hill Notation):
C30H50O3
CAS Number:
Molecular Weight:
458.72

Products

Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing