Skip to Content
MilliporeSigma
Sodium trifluoromethanesulfonimide 97%

Sodium trifluoromethanesulfonimide

Sodium trifluoromethanesulfonimide 97%
Synonym(s):
Sodium bis(trifluoromethylsulfonyl)imide
Empirical Formula (Hill Notation):
C2F6NNaO4S2
CAS Number:
Molecular Weight:
303.14

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing