Skip to Content
MilliporeSigma
Sodium triflinate ≥95.0% (T)

Sodium triflinate

Sodium triflinate ≥95.0% (T)
Synonym(s):
Langlois reagent, Sodium trifluoromethanesulfinate, Sodium trifluoromethylsulfinate, Trifluoromethanesulfinic acid sodium salt
Empirical Formula (Hill Notation):
CF3NaO2S
CAS Number:
Molecular Weight:
156.06

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing