Skip to Content
MilliporeSigma
Sodium tetrakis(4-fluorophenyl)borate dihydrate Selectophore™, ≥97.0%

Sodium tetrakis(4-fluorophenyl)borate dihydrate

Sodium tetrakis(4-fluorophenyl)borate dihydrate Selectophore™, ≥97.0%
Synonym(s):
Cesibor, Tetrakis(4-fluorophenyl)boron sodium
Linear Formula:
(FC6H4)4BNa·2H2O
CAS Number:
Molecular Weight:
450.21

Products

Product Category
Brand
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Reagent Type
Core
Special Grade