Skip to Content
MilliporeSigma
Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate Selectophore™

Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate

Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate Selectophore™
Synonym(s):
NaBARF, Tetrakis[3,5-bis(trifluoromethyl)phenyl]boron sodium
Empirical Formula (Hill Notation):
C32H12BF24Na
CAS Number:
Molecular Weight:
886.20

Products

Product Category
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Reagent Type
Core
Showing 1-2 of 2