Skip to Content
MilliporeSigma
Sodium (R)-β-hydroxyisobutyrate ≥96.0%

Sodium (R)-β-hydroxyisobutyrate

Sodium (R)-β-hydroxyisobutyrate ≥96.0%
Synonym(s):
(R)-β-Hydroxyisobutyric acid sodium salt, (R)-3-Hydroxy-2-methylpropionic acid sodium salt, R-HIBA, R-β-HIBA-Na, Sodium (R)-3-hydroxy-2-methylpropionate
Empirical Formula (Hill Notation):
C4H7NaO3
CAS Number:
Molecular Weight:
126.09

Products

Product Category
Brand
Formula Weight
Purity
Chemical Composition
Showing 1-1 of 1
Compare
Product Number
Description
Pricing