Skip to Content
MilliporeSigma
Sodium pyrophosphate tetrabasic decahydrate BioUltra, ≥99.5% (T)

Sodium pyrophosphate tetrabasic decahydrate

Sodium pyrophosphate tetrabasic decahydrate BioUltra, ≥99.5% (T)
Synonym(s):
Pyrophosphate inorganic, Sodium diphosphate tetrabasic, Tetrasodium pyrophosphate
Linear Formula:
Na4P2O7 · 10H2O
CAS Number:
Molecular Weight:
446.06
EC Number:

Products

Product Category
Brand
Feature
Formula Weight
pH Value
Physical Form
Product Line
Purity
Quality Segment