Skip to Content
MilliporeSigma
Sodium pyrophosphate decahydrate ACS reagent, ≥99%

Sodium pyrophosphate decahydrate

Sodium pyrophosphate decahydrate ACS reagent, ≥99%
Synonym(s):
Sodium pyrophosphate tetrabasic decahydrate, Tetrasodium pyrophosphate
Linear Formula:
Na4P2O7 · 10H2O
CAS Number:
Molecular Weight:
446.06
EC Number:

Products

Product Category
Brand
Formula Weight
pH Value
Physical Form
Purity
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing