Skip to Content
MilliporeSigma
Sodium polytungstate solution ~85% in H2O

Sodium polytungstate solution

Sodium polytungstate solution ~85% in H2O
Empirical Formula (Hill Notation):
Na6O39W12
CAS Number:
Molecular Weight:
2968.00

Products

Product Category
Color
Formula Weight
Physical Form
Chemical Composition
Application
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing