Skip to Content
MilliporeSigma
Sodium phosphate monobasic solution BioUltra, 5 M in H2O

Sodium phosphate monobasic solution

Sodium phosphate monobasic solution BioUltra, 5 M in H2O
Empirical Formula (Hill Notation):
H2NaO4P
CAS Number:
Molecular Weight:
119.98

Products

Product Category
Formula Weight
pH Value
Physical Form
Product Line
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing