Skip to Content
MilliporeSigma
Sodium phosphate monobasic suitable for HPLC, 99.0-101.0% (T)

Sodium phosphate monobasic

Sodium phosphate monobasic suitable for HPLC, 99.0-101.0% (T)
Synonym(s):
Monosodium dihydrogen orthophosphate, Monosodium phosphate, Sodium dihydrogen phosphate
Linear Formula:
NaH2PO4
CAS Number:
Molecular Weight:
119.98

Products

Product Category
Feature
Formula Weight
pH Value
Physical Form
pK Value
Product Line
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Technique