Skip to Content
MilliporeSigma
Sodium phenyl phosphate dibasic dihydrate ≥95%

Sodium phenyl phosphate dibasic dihydrate

Sodium phenyl phosphate dibasic dihydrate ≥95%
Synonym(s):
Disodium phenyl phosphate dihydrate, Phenyl phosphate disodium salt
Linear Formula:
C6H5PO4Na2 · 2H2O
CAS Number:
Molecular Weight:
254.09
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing