Skip to Content
MilliporeSigma
Sodium perchlorate hydrate 99.99% trace metals basis

Sodium perchlorate hydrate

Sodium perchlorate hydrate 99.99% trace metals basis
Linear Formula:
NaClO4 · xH2O
Molecular Weight:
122.44 (anhydrous basis)
EC Number:

Products

Formula Weight
Physical Form
Purity
Chemical Composition
Quality Segment
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing