Skip to Content
MilliporeSigma
Sodium nitrate BioXtra, ≥99.0%

Sodium nitrate

Sodium nitrate BioXtra, ≥99.0%
Synonym(s):
Chile salpeter, Sodium nitrate
Empirical Formula (Hill Notation):
NaNO3
CAS Number:
Molecular Weight:
84.99
EC Number:

Products

Product Category
Brand
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Technique