Skip to Content
MilliporeSigma
Sodium L-lactate-13C3 solution endotoxin tested, 45-55 % (w/w) in H2O, ≥99 atom % 13C, ≥98% (CP)

Sodium L-lactate-13C3 solution

Sodium L-lactate-13C3 solution endotoxin tested, 45-55 % (w/w) in H2O, ≥99 atom % 13C, ≥98% (CP)
Synonym(s):
L-Lactic acid-13C3 sodium salt solution
Linear Formula:
13CH313CH(OH)13CO2Na
CAS Number:
Molecular Weight:
115.04

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Isotope
Markush Class
Markush Group
Optical Isomer
Technique