Skip to Content
MilliporeSigma
Sodium iminodiacetate dibasic hydrate 98%

Sodium iminodiacetate dibasic hydrate

Sodium iminodiacetate dibasic hydrate 98%
Synonym(s):
Iminodiacetic acid disodium salt hydrate
Linear Formula:
C4H5NO4Na2 · xH2O
CAS Number:
Molecular Weight:
177.07 (anhydrous basis)

Products

Product Category
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing