Skip to Content
MilliporeSigma
Sodium hydrogen sulfate monohydrate extra pure, Technipur®

Sodium hydrogen sulfate monohydrate

Sodium hydrogen sulfate monohydrate extra pure, Technipur®
Synonym(s):
Sodium bisulfate monohydrate, Sodium hydrogen sulfate monohydrate
Linear Formula:
NaHSO4 · H2O
CAS Number:
Molecular Weight:
138.08

Products

Brand
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-2 of 2