Skip to Content
MilliporeSigma
Sodium hexafluoroaluminate 99.98% trace metals basis

Sodium hexafluoroaluminate

Sodium hexafluoroaluminate 99.98% trace metals basis
Synonym(s):
Cryolite, Cryolite, synthetic, Sodium aluminum hexafluoride
Linear Formula:
Na3AlF6
CAS Number:
Molecular Weight:
209.94
EC Number:

Products

Brand
Formula Weight
Physical Form
Purity
Reagent Type
Core
Showing 1-2 of 2
Compare
Product Number
Description
Pricing