Skip to Content
MilliporeSigma
Sodium glycodeoxycholate BioXtra, ≥97% (HPLC)

Sodium glycodeoxycholate

Sodium glycodeoxycholate BioXtra, ≥97% (HPLC)
Synonym(s):
3α,12α-Dihydroxy-5β-cholan-24-oic acid N-(carboxymethyl)amide, Glycodeoxycholic acid sodium salt, Glycodesoxycholic acid, N-(3α,12α-Dihydroxy-24-oxocholan-24-yl)glycine
Empirical Formula (Hill Notation):
C26H42NNaO5
CAS Number:
Molecular Weight:
471.61
EC Number:

Products

Product Category
Formula Weight
Product Line
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing