Skip to Content
MilliporeSigma
Sodium glycocholate hydrate ≥95% (TLC)

Sodium glycocholate hydrate

Sodium glycocholate hydrate ≥95% (TLC)
Synonym(s):
3α,7α,12α-Trihydroxy-5β-cholan-24-oic acid N-(carboxymethyl)amide sodium salt, Glycocholic acid sodium salt hydrate, N-(3α,7α,12α-Trihydroxy-24-oxocholan-24-yl)glycine sodium salt, N-Cholylglycine sodium salt
Linear Formula:
C26H42NO6Na · xH2O
CAS Number:
Molecular Weight:
487.60 (anhydrous basis)
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing