Skip to Content
MilliporeSigma
Sodium fumarate dibasic ≥99%

Sodium fumarate dibasic

Sodium fumarate dibasic ≥99%
Synonym(s):
Disodium fumarate, Fumaric acid disodium salt
Linear Formula:
NaOOCCH=CHCOONa
CAS Number:
Molecular Weight:
160.04
EC Number:

Products

Product Category
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing