Skip to Content
MilliporeSigma
Sodium benzoate certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sodium benzoate

Sodium benzoate certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
Sodium benzoate, Benzoic acid sodium salt
Linear Formula:
C6H5COONa
CAS Number:
Molecular Weight:
144.10
EC Number:

Products

Product Category
Brand
Feature
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique