Skip to Content
MilliporeSigma
Sodium arsenate dibasic heptahydrate ≥98.0%

Sodium arsenate dibasic heptahydrate

Sodium arsenate dibasic heptahydrate ≥98.0%
Synonym(s):
Disodium hydrogen arsenate heptahydrate, di-Sodium hydrogen arsenate heptahydrate
Linear Formula:
Na2HAsO4 · 7H2O
CAS Number:
Molecular Weight:
312.01

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Reagent Type
Core
Special Grade
Showing 1-2 of 2