Skip to Content
MilliporeSigma
Sodium acetate solution BioUltra, Molecular Biology, ~3 M in H2O

Sodium acetate solution

Sodium acetate solution BioUltra, Molecular Biology, ~3 M in H2O
Empirical Formula (Hill Notation):
C2H3NaO2
CAS Number:
Molecular Weight:
82.03

Products

Product Category
Brand
Formula Weight
pH Value
Physical Form
Product Line
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing