Skip to Content
MilliporeSigma
Sodium 4-hydroxybenzenesulfonate dihydrate 98%

Sodium 4-hydroxybenzenesulfonate dihydrate

Sodium 4-hydroxybenzenesulfonate dihydrate 98%
Synonym(s):
4-Hydroxybenzenesulfonic acid sodium salt, Phenol-4-sulfonic acid sodium salt dihydrate
Linear Formula:
HOC6H4SO3Na·2H2O
CAS Number:
Molecular Weight:
232.19
EC Number:

Products

Product Category
Brand
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing