Skip to Content
MilliporeSigma
Sodium 3-sulfobenzoate 97%

Sodium 3-sulfobenzoate

Sodium 3-sulfobenzoate 97%
Synonym(s):
3-Carboxybenzenesulfonic acid sodium salt, 3-Sulfobenzoic acid sodium salt
Linear Formula:
NaO3SC6H4CO2H
CAS Number:
Molecular Weight:
224.17
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing