Skip to Content
MilliporeSigma
Sodium 3,5-dichloro-2-hydroxybenzenesulfonate used for peroxide measurement

Sodium 3,5-dichloro-2-hydroxybenzenesulfonate

Sodium 3,5-dichloro-2-hydroxybenzenesulfonate used for peroxide measurement
Synonym(s):
3,5-Dichloro-2-hydroxybenzenesulfonic acid, DCHBS, DHBS
Empirical Formula (Hill Notation):
C6H3Cl2NaO4S
CAS Number:
Molecular Weight:
265.05
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Application
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing