Skip to Content
MilliporeSigma
Sodium 2-ethylhexyl sulfate ~50% in H2O

Sodium 2-ethylhexyl sulfate

Sodium 2-ethylhexyl sulfate ~50% in H2O
Synonym(s):
2-Ethylhexyl sulfate sodium salt
Empirical Formula (Hill Notation):
C8H17NaO4S
CAS Number:
Molecular Weight:
232.27

Products

Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing