Skip to Content
MilliporeSigma
Skatole ≥98%

Skatole

Skatole ≥98%
Synonym(s):
3-Methylindole, Skatole
Empirical Formula (Hill Notation):
C9H9N
CAS Number:
Molecular Weight:
131.17
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Agency
Application
Markush Class
Markush Group
Organoleptic Class
Showing 1-1 of 1
Compare
Product Number
Description
Pricing