Skip to Content
MilliporeSigma
Silver (I) Acetate anhydrous, ≥99.9% trace metals basis

Silver (I) Acetate

Silver (I) Acetate anhydrous, ≥99.9% trace metals basis
Synonym(s):
Silver acetate, Acetic acid silver salt
Linear Formula:
CH3COOAg
CAS Number:
Molecular Weight:
166.91

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing