SILu™MAb K4 Stable-Isotope Labeled Universal Monoclonal Antibody
Synonym(s):
SILu™MAB Stable-Isotope Labeled Universal Monoclonal Antibody Standard human
Products
Product Category
Brand
Purity
Recombinant Host
Quality Segment
Package Size
Showing 1-1 of 1
Compare
Product Number
Description
Pricing