Skip to Content
MilliporeSigma
Silthiofam PESTANAL®, analytical standard

Silthiofam

Silthiofam PESTANAL®, analytical standard
Synonym(s):
N-Allyl-4,5-dimethyl-2-(trimethylsilyl)thiophene-3-carboxamide
Empirical Formula (Hill Notation):
C13H21NOSSi
CAS Number:
Molecular Weight:
267.46
EC Number:

Products

Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique