Skip to Content
MilliporeSigma
Silibinin ≥98% (HPLC)

Silibinin

Silibinin ≥98% (HPLC)
Synonym(s):
2,3-Dihydro-3-(4-hydroxy-3-methoxyphenyl)-2-(hydroxymethyl)-6-(3,5,7-trihydroxy-4-oxobenzopyran-2-yl)benzodioxin, Silybin
Empirical Formula (Hill Notation):
C25H22O10
CAS Number:
Molecular Weight:
482.44
EC Number:

Products

Brand
Formula Weight
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing