Skip to Content
MilliporeSigma
Sibutramine hydrochloride solution 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sibutramine hydrochloride solution

Sibutramine hydrochloride solution 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Synonym(s):
Sibutramine hydrochloride monohydrate, 1-(4-Chlorophenyl)-N,N-dimethyl-alpha-(2-methylpropyl)cyclobutanemethanamine hydrochloride monohydrate, Meridia, Reductil
Empirical Formula (Hill Notation):
C17H26ClN · HCl
CAS Number:
Molecular Weight:
316.31
EC Number:

Products

Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Application
Package Size
Reference Material Type
Special Grade
Technique