![S-Hydroprene PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/207/887/733043d5-4bed-44f3-b1c8-432509c43f36/640/733043d5-4bed-44f3-b1c8-432509c43f36.png)
S-Hydroprene
![S-Hydroprene PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/207/887/733043d5-4bed-44f3-b1c8-432509c43f36/640/733043d5-4bed-44f3-b1c8-432509c43f36.png)
Synonym(s):
Ethyl 3,7,11-trimethyl-2,4-dodecadienoate
Empirical Formula (Hill Notation):
C17H30O2
CAS Number:
Molecular Weight:
266.42
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing