Skip to Content
MilliporeSigma
Shi Epoxidation Diketal Catalyst 98%

Shi Epoxidation Diketal Catalyst

Shi Epoxidation Diketal Catalyst 98%
Synonym(s):
1,2:4,5-Di-O-isopropylidene-β-D-erythro-2,3-hexodiulo-2,6-pyranose
Empirical Formula (Hill Notation):
C12H18O6
CAS Number:
Molecular Weight:
258.27

Products

Product Category
Sustainability
Feature
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing