Skip to Content
MilliporeSigma
Sertraline hydrochloride impurity standard British Pharmacopoeia (BP) Reference Standard

Sertraline hydrochloride impurity standard

Sertraline hydrochloride impurity standard British Pharmacopoeia (BP) Reference Standard
Synonym(s):
Sertraline hydrochloride, (1S,4S)-4-(3,4-Dichlorophenyl)-1,2,3,4-tetrahydro-N-methyl-1-naphthalenamine hydrochloride
Empirical Formula (Hill Notation):
C17H17NCl2 · HCl
CAS Number:
Molecular Weight:
342.69

Products

Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1