Skip to Content
MilliporeSigma
Semicarbazide hydrochloride VETRANAL®, analytical standard

Semicarbazide hydrochloride

Semicarbazide hydrochloride VETRANAL®, analytical standard
Synonym(s):
N-Aminourea hydrochloride, Hydrazine carboxamide hydrochloride
Linear Formula:
NH2CONHNH2 · HCl
CAS Number:
Molecular Weight:
111.53
EC Number:

Products

Product Category
Formula Weight
Melting Point (°C)
Product Line
Purity
Format
Research Area
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing